Calcium chloride 90-98%, TECHNICAL
Supplier: VWR Chemicals
Warning
Synonyms: Chloro calcium, Calcium dichloride, CaCl2
22313.294EA 104 EUR
22313.294 22313.363 22313.460
Calcium chloride 90-98%, TECHNICAL
Calcium chloride
Formula: CaCl₂
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 772 °C
Density: 2.174 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659
Order Now
Specification Test Results
Assay 90 - 98 %