Acetylenedicarboxylic acid 98%
Supplier: Thermo Fisher Scientific
Danger
102520250 102521000
ACRO102520250EA 136 EUR
ACRO102520250 ACRO102521000
Acetylenedicarboxylic acid 98%
Acetylenedicarboxylic acid
Formula: C₄H₂O₄
MW: 114.06 g/mol
Melting Pt: 180…187 °C (dec.)
Flash Pt: 131 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004362
CAS Number: 142-45-0
EINECS: 205-536-0
UN: 2811
ADR: 6.1 III
Order Now
Specification Test Results
Appearance White to cream to beige Crystalline powder or crystals
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
GC ≥98.5 %