Diisopropyl phosphonate 98%
Supplier: Thermo Scientific
ACRO115261000EA 125 EUR
ACRO115261000
Diisopropyl phosphonate 98%
Diisopropyl phosphonate
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4065 to 1.4085 (20°C, 589 nm)
GC ≥97.5 %