Bis(cyclopentadienyl)zirconium dichloride 98+%
Supplier: Thermo Scientific
Warning
Synonyms: Zirconocene dichloride
187920050 187920250 187921000
ACRO187920050EA 46.9 EUR
ACRO187920050 ACRO187920250 ACRO187921000
Bis(cyclopentadienyl)zirconium dichloride 98+%
Bis(cyclopentadienyl)zirconium dichloride
Formula: C₁₀H₁₀Cl₂Zr
MW: 292.32 g/mol
Melting Pt: 242…245 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003726
CAS Number: 1291-32-3
EINECS: 215-066-8
ADR: 8,III
Order Now
Specification Test Results
Appearance White to light beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 242 to 245 °C
Titration Argentometric ≥98.0 %