Platinum 10% on activated carbon wetted with ca. 50% water hydrogenation catalyst
Supplier: Thermo Fisher Scientific
Synonyms: Platinium
Stabilization: moistured with water
195240100 195240500 195240010
ACRO195240100EA 201 EUR
ACRO195240100 ACRO195240500 ACRO195240010
Platinum 10% on activated carbon wetted with ca. 50% water hydrogenation catalyst
Platinum
Formula: Pt
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
ADR: 4.2,III
Order Now
Specification Test Results
Appearance Black Powder
Moisture 45 % to 60 %
Platinum (Pt) 9.8 to 10.2 %