3-Hydroxypicolinic acid 98%
Supplier: Thermo Scientific
Warning
Synonyms: 3-Hydroxypyridine-2-carboxylic acid
241050050 241050250
ACRO241050050EA 118 EUR
ACRO241050050 ACRO241050250
3-Hydroxypicolinic acid 98%
3-Hydroxypicolinic acid
Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 208…212 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006294
CAS Number: 874-24-8
EINECS: 212-859-0
Order Now
Specification Test Results
Appearance Beige to brown Powder
Infrared spectrum Conforms
Melting point 208 to 214 °C
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %