N-Methylpropylamine 96%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: N-Methyl-1-propylamine
314690050 314690250
ACRO314690050EA 68.8 EUR
ACRO314690050 ACRO314690250
N-Methylpropylamine 96%
N-Methylpropylamine
Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 61…63 °C (760 mmHg)
Density: 0.71 g/cm³
Flash Pt: <–34 °C
Storage Temperature: Ambient
MDL Number: MFCD00009361
CAS Number: 627-35-0
UN: 2733
ADR: 3 II
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3890 to 1.3940 (20°C, 589 nm)
GC ≥95 %