Bis(cyclopentadienyl)titanium dichloride ≥97%
Supplier: Thermo Scientific
Warning
Synonyms: Titanocenedichloride, Bis(cyclopentadienyl)titanium(IV) dichloride, Titanocene dichloride
A11456.09 A11456.18 A11456.30
A11456.09EA 48.1 EUR
A11456.09 A11456.18 A11456.30
Bis(cyclopentadienyl)titanium dichloride ≥97%
Bis(cyclopentadienyl)titanium dichloride
Purity: 97% CAS No.: 1271-19-8 Molecular Formula: C10H10Cl2Ti Formula Weight: 249.00 Odour: Odourless Melting Point: ca 285° dec. MDL No.: MFCD00003723
Formula: C₁₀H₁₀Cl₂Ti
MW: 248.97 g/mol
Melting Pt: ∼ 285 °C (dec.)
Density: 1.60
Storage Temperature: Ambient
MDL Number: MFCD00003723
CAS Number: 1271-19-8
EINECS: 215-035-9
UN: 3261
ADR: 8,II
Merck Index: 14,9482