Benzoyl ferrocene ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: Benzoylferrocene

A17685.06 A17685.14 A17685.03
A17685.06EA 48 EUR
A17685.06 A17685.14 A17685.03
Benzoyl ferrocene ≥98%
Benzoylferrocene

Purity: 98% CAS No.: 1272-44-2 Molecular Formula: C17H14FeO Formula Weight: 290.14 Melting Point: 106-109° MDL No.: MFCD00001431

Formula: C₁₂H₁₂FeO
MW: 290.14 g/mol
Melting Pt: 106…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00001431
CAS Number: 1272-44-2
EINECS: 215-054-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR