2-(2,4-Difluorophenoxy)-3-nitropyridine
Supplier: Thermo Scientific
BTB08013.1 BTB08013.10
MAYBBTB08013.1EA 0 EUR
MAYBBTB08013.1 MAYBBTB08013.10
2-(2,4-Difluorophenoxy)-3-nitropyridine
2-(2,4-Difluorophenoxy)-3-nitropyridine
Formula: C₁₁H₆F₂N₂O₃
MW: 252.18 g/mol
MDL Number: MFCD00067800
CAS Number: 175135-62-3