Ethyl-5-bromonicotinate

Supplier: Thermo Scientific

Synonyms: Ethyl 5-bromonicotinate

RDP00009.1 RDP00009.10 RDP00009.50
MAYBRDP00009.1EA 0 EUR
MAYBRDP00009.1 MAYBRDP00009.10 MAYBRDP00009.50
Ethyl-5-bromonicotinate
Ethyl 5-bromonicotinate
Formula: C₈H₈BrNO₂
MW: 230.06 g/mol
MDL Number: MFCD00040366
CAS Number: 20986-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR