Methyl 2,6-dichloro-5-fluoronicotinate
Supplier: Thermo Scientific
Synonyms:
Methyl 2,6-dichloro-5-fluoronicotinate
RH02135DA
RH02135EA
MAYBRH02135DAEA
0
EUR
MAYBRH02135DA
MAYBRH02135EA
Methyl 2,6-dichloro-5-fluoronicotinate
Methyl 2,6-dichloro-5-fluoronicotinate
Formula:
C₇H₄Cl₂FNO₂ MW: 224.02 g/mol |
MDL Number:
MFCD02180634 CAS Number: 189281-66-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...