4-Chlorobenzylidenemalononitrile
Supplier: Thermo Scientific
Synonyms:
2-(4-Chlorobenzylidene)malononitrile
S02458.250
S02458.1
MAYBS02458.250EA
0
EUR
MAYBS02458.250
MAYBS02458.1
4-Chlorobenzylidenemalononitrile
4-Chlorobenzylidenemalononitrile
Formula:
C₁₀H₅ClN₂ MW: 188.62 g/mol |
MDL Number:
MFCD00126183 CAS Number: 1867-38-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...