Malachite green oxalate
Supplier: Roth Carl
Synonyms:
Bis (N-{4-[[4-(dimethylamino)phenyl](phenyl)methylene]cyclohexa-2,5-dien-1-ylidene}-N-methylmethanaminium) trioxalate
, Basic Green 4
, N,N,N',N'-Tetramethyl-4,4'-diaminotriphenylcarbenium oxalate
ROTH5895.2EA
21.2
EUR
ROTH5895.2
Malachite green oxalate
Malachite green oxalate
Formula:
C₅₂H₅₄N₄O₁₂ MW: 927,02 g/mol Melting Pt: ∼159 °C |
MDL Number:
MFCD00011766 CAS Number: 2437-29-8 UN: 2811 ADR: 6.1,III Merck Index: 13,05722 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...