Cyanoacetamide, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-Cyanoacetamide

M-Clarity™ quality level = MQ200

108448-100G 108448-500G
SIAL108448-100GEA 54.5 EUR
SIAL108448-100G SIAL108448-500G
Cyanoacetamide, Sigma-Aldrich®
Cyanoacetamide
Formula: C₃H₄N₂O
MW: 84.08 g/mol
MDL Number: MFCD00008024
CAS Number: 107-91-5
EINECS: 203-531-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR