Sodium benzenesulphinate, Sigma-Aldrich®
Supplier: Merck
Synonyms: Benzenesulphinic acid sodium salt, Benzenesulphinic acid, sodium salt
123579-100G 123579-25G 123579-500G
SIAL123579-100GEA 60.1 EUR
SIAL123579-100G SIAL123579-25G SIAL123579-500G
Sodium benzenesulphinate, Sigma-Aldrich®
Sodium benzenesulphinate
Formula: C₆H₅NaO₂S
MW: 164.16 g/mol
MDL Number: MFCD00013135
CAS Number: 873-55-2
EINECS: 212-842-8