Methacrolein stabilised, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Acrolein
Stabilization:
Stabilised with hydroquinone (HQ)
M-Clarity™ quality level = MQ100
133035-25ML
133035-5ML
SIAL133035-25MLEA
173
EUR
SIAL133035-25ML
SIAL133035-5ML
Methacrolein stabilised, Sigma-Aldrich®
Methacrolein
Formula:
C₄H₆O MW: 70.09 g/mol |
MDL Number:
MFCD00006974 CAS Number: 78-85-3 EINECS: 201-150-1 UN: 2396 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...