Isobutyl methacrylate stabilised, Sigma-Aldrich®

Supplier: Merck

Synonyms: Methacrylic acid Isobutyl ester

Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200

169919-18L 169919-1L
SIAL169919-18LEA 690 EUR
SIAL169919-18L SIAL169919-1L
Isobutyl methacrylate stabilised, Sigma-Aldrich®
Isobutyl methacrylate
Formula: C₈H₁₄O₂
MW: 142.2 g/mol
MDL Number: MFCD00008931
CAS Number: 97-86-9
EINECS: 202-613-0
UN: 2283
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR