Iodosobenzene diacetate, Sigma-Aldrich®

Supplier: Merck

Synonyms: Iodobenzene diacetate, (Diacetoxyiodo)benzene

M-Clarity™ quality level = MQ200

178721-100G 178721-1KG 178721-25G 178721-5G
SIAL178721-100GEA 203 EUR
SIAL178721-100G SIAL178721-1KG SIAL178721-25G SIAL178721-5G
Iodosobenzene diacetate, Sigma-Aldrich®
Iodosobenzene diacetate
Formula: C₁₀H₁₁IO₄
MW: 322.1 g/mol
MDL Number: MFCD00008692
CAS Number: 3240-34-4
EINECS: 221-808-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR