4-Nitrobenzyl chloroformate, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Nitrobenzyl carbonochloridate

222801-25G 222801-5G
SIAL222801-25GEA 111 EUR
SIAL222801-25G SIAL222801-5G
4-Nitrobenzyl chloroformate, Sigma-Aldrich®
4-Nitrobenzyl chloroformate
Formula: C₈H₆ClNO₄
MW: 215.59 g/mol
MDL Number: MFCD00007375
CAS Number: 4457-32-3
EINECS: 224-708-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR