N,N-Dimethylacetamide dimethyl acetal, Sigma-Aldrich®

Supplier: Merck

Synonyms: (1,1-Dimethoxyethyl)dimethylamine, 1,1-Dimethoxy-N,N-dimethylethanamine

261483-25G 261483-5G
SIAL261483-25GEA 260 EUR
SIAL261483-25G SIAL261483-5G
N,N-Dimethylacetamide dimethyl acetal, Sigma-Aldrich®
N,N-Dimethylacetamide dimethyl acetal
Formula: C₆H₁₅NO₂
MW: 133.19 g/mol
MDL Number: MFCD00008476
CAS Number: 18871-66-4
EINECS: 242-641-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR