Crotonaldehyde, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Butenal, mixture of cis and trans, Crotonaldehyde, mixture of cis and trans
MQ200
262668-100ML 262668-1L 262668-5ML
SIAL262668-100MLEA 74.5 EUR
SIAL262668-100ML SIAL262668-1L SIAL262668-5ML
Crotonaldehyde, Sigma-Aldrich®
Crotonaldehyde
Formula: C₄H₆O
MW: 70.09 g/mol
MDL Number: MFCD00007003
CAS Number: 4170-30-3
EINECS: 224-030-0
UN: 1143
ADR: 6.1 I