2-Hydroxypropyl methacrylate (mixture of isomers) ≥97% stabilised, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Propylene Glycol Monomethacrylate, HPMA, Hydroxypropyl methacrylate, Methacrylic acid 2-hydroxypropyl ester, Methacrylic acid hydroxypropyl ester
Stabilization: Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) 180 - 220 ppm
M-Clarity™ quality level = MQ200
268542-100ML 268542-18L 268542-1L
SIAL268542-100MLEA 54.1 EUR
SIAL268542-100ML SIAL268542-18L SIAL268542-1L
2-Hydroxypropyl methacrylate (mixture of isomers) ≥97% stabilised, Sigma-Aldrich®
2-Hydroxypropyl methacrylate (mixture of isomers, methacrylic acid, monoester with propane-1,2-diol)
Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Flash Pt: 95 °C
MDL Number: MFCD00004536
CAS Number: 27813-02-1
EINECS: 248-666-3