(S)-(+)-β-Aminophenethyl alcohol, Sigma-Aldrich®

Supplier: Merck

Synonyms: (S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol

282693-1G 282693-5G
SIAL282693-1GEA 27.2 EUR
SIAL282693-1G SIAL282693-5G
(S)-(+)-β-Aminophenethyl alcohol, Sigma-Aldrich®
(S)-(+)-β-Aminophenethyl alcohol
Formula: C₈H₁₁NO
MW: 137.18 g/mol
MDL Number: MFCD00064404
CAS Number: 20989-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR