N,N-Dimethyl-1,3-diaminopropane, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 3-Dimethylaminopropylamine, N,N-Dimethyltrimethylenediamine, N,N-Dimethyl-1,3-propanediamine

M-Clarity™ quality level = MQ200

39380-1L 39380-250ML
SIAL39380-1LEA 63.8 EUR
SIAL39380-1L SIAL39380-250ML
N,N-Dimethyl-1,3-diaminopropane, Sigma-Aldrich®
N,N-Dimethyl-1,3-diaminopropane
Formula: C₅H₁₄N₂
MDL Number: MFCD00008216
CAS Number: 109-55-7
EINECS: 203-680-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR