Tetrafluoroboric acid diethyl ether complex (51 - 57% HBF₄) (w/w), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
400068-100ML 400068-25ML 400068-400ML
SIAL400068-100MLEA 131 EUR
SIAL400068-100ML SIAL400068-25ML SIAL400068-400ML
Tetrafluoroboric acid diethyl ether complex (51 - 57% HBF₄) (w/w), Sigma-Aldrich®
Tetrafluoroboric acid-diethyl ether complex (50-55% tetrafluoroboric acid in diethyl ether)
Formula: C₄H₁₁BF₄O
MW: 161.93 g/mol
MDL Number: MFCD00085352
CAS Number: 67969-82-8
EINECS: 267-983-8
UN: 3129
ADR: 4.3 (8) II