1,4-Butanediol diacrylate stabilised, Sigma-Aldrich®
Supplier: Merck
Stabilization: Stabilised with hydroquinone (HQ)
M-Clarity™ quality level = MQ200
411744-100ML 411744-25ML
SIAL411744-100MLEA 63.8 EUR
SIAL411744-100ML SIAL411744-25ML
1,4-Butanediol diacrylate stabilised, Sigma-Aldrich®
1,4-Butanediol diacrylate
Formula: C₁₀H₁₄O₄
MW: 198.22 g/mol
MDL Number: MFCD00014940
CAS Number: 1070-70-8
EINECS: 213-979-6