Acryloyl chloride stabilised, Sigma-Aldrich®
Supplier: Merck
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ)
M-Clarity™ quality level = MQ200
549797-500G
549797-5G
SIAL549797-500GEA
990
EUR
SIAL549797-500G
SIAL549797-5G
Acryloyl chloride stabilised, Sigma-Aldrich®
Acryloyl chloride
Formula:
C₃H₃ClO MW: 90.51 g/mol |
MDL Number:
MFCD00000731 CAS Number: 814-68-6 EINECS: 212-399-0 UN: 3383 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...