5-Bromo-2-hydroxy-3-nitropyridine, Sigma-Aldrich®

Supplier: Merck

Synonyms: 5-Bromo-3-nitro-2-pyridinol, 5-Bromo-3-nitropyridin-2(1H)-one

658448-25G 658448-5G
SIAL658448-25GEA 151 EUR
SIAL658448-25G SIAL658448-5G
5-Bromo-2-hydroxy-3-nitropyridine, Sigma-Aldrich®
5-Bromo-2-hydroxy-3-nitropyridine
Formula: C₅H₃BrN₂O₃
MW: 218.99 g/mol
MDL Number: MFCD00023473
CAS Number: 15862-34-7
EINECS: 239-989-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR