(S)-(+)-3,3-Dimethyl-2-butylamine, Sigma-Aldrich®
Supplier: Merck
Synonyms: (S)-3,3-Dimethylbutan-2-amine
M-Clarity™ quality level = MQ100
668516-25G 668516-5G
SIAL668516-25GEA 598 EUR
SIAL668516-25G SIAL668516-5G
(S)-(+)-3,3-Dimethyl-2-butylamine, Sigma-Aldrich®
(S)-(+)-3,3-Dimethyl-2-butylamine
Formula: C₆H₁₅N
MW: 101.19 g/mol
MDL Number: MFCD00671644
CAS Number: 22526-47-2