1,3-Diiodo-5,5-dimethylhydantoin, Sigma-Aldrich®
Supplier: Merck
Synonyms:
DIH
711624-25G
711624-5G
SIAL711624-25GEA
458
EUR
SIAL711624-25G
SIAL711624-5G
1,3-Diiodo-5,5-dimethylhydantoin, Sigma-Aldrich®
1,3-Diiodo-5,5-dimethylhydantoin
Formula:
C₅H₆I₂N₂O₂ MW: 379.92 g/mol |
MDL Number:
MFCD00020867 CAS Number: 2232-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...