Platinum on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Supplier: Merck
Synonyms: Platinium
80980-1G 80980-5G
SIAL80980-1GEA 58.1 EUR
SIAL80980-1G SIAL80980-5G
Platinum on activated carbon hydrogenation catalyst, Sigma-Aldrich®
Platinum
Formula: Pt
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 3088
ADR: 4.2,III