5,5-Dimethylhydantoin, Sigma-Aldrich®
Supplier: Merck
Synonyms: 5,5-Dimethylimidazolidine-2,4-dione
M-Clarity™ quality level = MQ200
D161403-100G D161403-1KG
SIALD161403-100GEA 28.1 EUR
SIALD161403-100G SIALD161403-1KG
5,5-Dimethylhydantoin, Sigma-Aldrich®
5,5-Dimethylhydantoin
Formula: C₅H₈N₂O₂
MW: 128.13 g/mol
MDL Number: MFCD00005266
CAS Number: 77-71-4
EINECS: 201-051-3