N,N-Dimethylsulphamoyl chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Dimethylsulphamoyl chloride
M-Clarity™ quality level = MQ200
D186252-100G D186252-25G D186252-500G
SIALD186252-100GEA 372 EUR
SIALD186252-100G SIALD186252-25G SIALD186252-500G
N,N-Dimethylsulphamoyl chloride, Sigma-Aldrich®
N,N-Dimethylsulphamoyl chloride
Formula: C₂H₆ClNO₂S
MW: 143.59 g/mol
MDL Number: MFCD00007425
CAS Number: 13360-57-1
EINECS: 236-412-4