O,O-Diethyl phosphorodithioate, Sigma-Aldrich®
Supplier: Merck
Synonyms:
O,O-Diethyl dithiophosphate
M-Clarity™ quality level = MQ100
D93600-100G
D93600-500G
SIALD93600-100GEA
201
EUR
SIALD93600-100G
SIALD93600-500G
O,O-Diethyl phosphorodithioate, Sigma-Aldrich®
O,O-Diethyl phosphorodithioate
Formula:
C₄H₁₁O₂PS₂ MW: 186.24 g/mol |
CAS Number:
298-06-6 EINECS: 206-055-9 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...