Isatoic anhydride, Sigma-Aldrich®

Supplier: Merck

Synonyms: 1H-Benzo[d][1,3]oxazine-2,4-dione

M-Clarity™ quality level = MQ100

I12808-100G I12808-500G
SIALI12808-100GEA 30.4 EUR
SIALI12808-100G SIALI12808-500G
Isatoic anhydride, Sigma-Aldrich®
Isatoic anhydride
Formula: C₈H₅NO₃
MW: 163.13 g/mol
MDL Number: MFCD00006700
CAS Number: 118-48-9
EINECS: 204-255-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR