Succinyl chloride, Sigma-Aldrich®

Supplier: Merck

Synonyms: Succinyl dichloride

M-Clarity™ quality level = MQ200

S6452-100G S6452-25G
SIALS6452-100GEA 317 EUR
SIALS6452-100G SIALS6452-25G
Succinyl chloride, Sigma-Aldrich®
Succinyl chloride
Formula: C₄H₄Cl₂O₂
MW: 154.98 g/mol
MDL Number: MFCD00000749
CAS Number: 543-20-4
EINECS: 208-838-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR