Benzylideneacetone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 4-Phenylbut-3-en-2-one, 4-Phenyl-3-buten-2-one

W288101-10KG-K W288101-1KG-K W288101-25KG-K
SIALW288101-10KG-KEA 576 EUR
SIALW288101-10KG-K SIALW288101-1KG-K SIALW288101-25KG-K
Benzylideneacetone, Sigma-Aldrich®
Benzylideneacetone
Formula: C₁₀H₁₀O
MW: 146.19 g/mol
MDL Number: MFCD00008779
CAS Number: 122-57-6
EINECS: 204-555-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR