Thallium(I) iodide ≥99.9995% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Thallium iodide
43001.09 43001.18 43001.36
43001.09EA 106 EUR
43001.09 43001.18 43001.36
Thallium(I) iodide ≥99.9995% (metals basis), Puratronic®
Thallium (I) iodide
Formula: ITl
MW: 331.29 g/mol
Boiling Pt: 824 °C
Melting Pt: 440
Density: 7.1
Storage Temperature: Ambient
MDL Number: MFCD00011279
CAS Number: 7790-30-9
EINECS: 232-199-7
UN: 1707
ADR: 6.1 II
Merck Index: 14,9264