Thallium(I) nitrate, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ200
309230-100G 309230-25G
SIAL309230-100GEA 317 EUR
SIAL309230-100G SIAL309230-25G
Thallium(I) nitrate, Sigma-Aldrich®
Formula: NO₃Tl
Melting Pt: 206 °C
Density: 1.273 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011280
CAS Number: 10102-45-1
EINECS: 233-273-1
UN: 2727
ADR: 6.1,II
Merck Index: 12,09400