Silica gel
Supplier: Roth Carl
T858.1 T858.2
ROTHT858.1EA 53.1 EUR
ROTHT858.1 ROTHT858.2
Silica gel
Silica gel
Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230
Melting Pt: 1710
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
REACH: 01-2119379499-16