Diphenyl diselenide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

SIAL42944-10GEA 89.3 EUR
SIAL42944-10G SIAL42944-50G
Diphenyl diselenide, Sigma-Aldrich®
Diphenyl diselenide
Formula: C₁₂H₁₀Se₂
MW: 312,13 g/mol
Boiling Pt: 377 °C (1013 hPa)
Melting Pt: 58…61 °C
Density: 1,32 g/cm³ (20 °C)
MDL Number: MFCD00003001
UN: 3283
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR