4-Bromoisoquinoline
Supplier: Thermo Scientific
|
Warning
|
CC04910.1
CC04910.10
CC04910.50
MAYBCC04910.1EA
0
EUR
MAYBCC04910.1
MAYBCC04910.10
MAYBCC04910.50
4-Bromoisoquinoline
4-Bromoisoquinoline
Formula:
C₉H₆BrN Boiling Pt: 280 °C (1013 hPa) Melting Pt: 42…42.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006904 CAS Number: 1532-97-4 EINECS: 216-244-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...