Isoquinoline, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
I28208-500G
I28208-100G
I28208-5G
SIALI28208-500GEA
206
EUR
SIALI28208-500G
SIALI28208-100G
SIALI28208-5G
Isoquinoline, Sigma-Aldrich®
Isoquinoline
Formula:
C₉H₇N MW: 129.16 g/mol Boiling Pt: 242 °C (1013 hPa) Melting Pt: 21…26 °C Density: 1.098 g/cm³ (20 °C) Flash Pt: 107 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006898 CAS Number: 119-65-3 EINECS: 204-341-8 UN: 2811 ADR: 6.1,II Merck Index: 13,05242 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...