Zinc oxide, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
204951-25G 204951-125G
SIAL204951-25GEA 163 EUR
SIAL204951-25G SIAL204951-125G
Zinc oxide, Sigma-Aldrich®
Zinc oxide
Formula: ZnO
Melting Pt: 1975 °C
Density: 5.61 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011300
CAS Number: 1314-13-2
EINECS: 215-222-5
UN: 3077
ADR: 9,III
REACH: 01-2119463881-32
Merck Index: 13,10200