1-Bromonaphthalene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: α-Bromonaphthalene
17640-100ML 17640-500ML
SIAL17640-100MLEA 120 EUR
SIAL17640-100ML SIAL17640-500ML
1-Bromonaphthalene, Sigma-Aldrich®
1-Bromonaphthalene
Formula: C₁₀H₇Br
MW: 207.07 g/mol
Boiling Pt: 280
Melting Pt: 0…6 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: 66
Storage Temperature: Ambient
MDL Number: MFCD00003868
CAS Number: 90-11-9
EINECS: 201-965-2
Merck Index: 13,01410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea9a18f77a334f3a7a688e768c8efc3a557bb3ab&vid=6a108e0806e706c6cdc50ad3d7fc0539d1d6a415&prid=web&clid=SAPDAM