1-Bromonaphthalene, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: α-Bromonaphthalene
M-Clarity™ quality level = MQ200
B73104-5G B73104-100G B73104-500G
SIALB73104-5GEA 48.2 EUR
SIALB73104-5G SIALB73104-100G SIALB73104-500G
1-Bromonaphthalene, Sigma-Aldrich®
1-Bromonaphthalene
Formula: C₁₀H₇Br
MW: 207.07 g/mol
Boiling Pt: 280 °C (1013 hPa)
Melting Pt: 0…6 °C
Density: 1.48 g/cm³ (20 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00003868
CAS Number: 90-11-9
EINECS: 201-965-2
Merck Index: 13,01410
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea9a18f77a334f3a7a688e768c8efc3a557bb3ab&vid=6a108e0806e706c6cdc50ad3d7fc0539d1d6a415&prid=web&clid=SAPDAM