1-Naphthol, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 1-Hydroxynaphthalene, α-Naphthol, α-Hydroxynaphthalene

M-Clarity™ quality level = MQ100

33420-100G-R 33420-250G-R
SIAL33420-100G-REA 188 EUR
SIAL33420-100G-R SIAL33420-250G-R
1-Naphthol, Sigma-Aldrich®
1-Naphthol
Formula: C₁₀H₈O
MW: 144.17 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 94 °C
Density: 1.224 g/cm³ (20 °C)
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00003930
CAS Number: 90-15-3
EINECS: 201-969-4
UN: 2811
ADR: 6.1,III
Merck Index: 13,06409

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR