Naphthalene, Sigma-Aldrich®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ100
184500-250G 184500-1KG 184500-5G
SIAL184500-250GEA 65.7 EUR
SIAL184500-250G SIAL184500-1KG SIAL184500-5G
Naphthalene, Sigma-Aldrich®
Naphthalene
Formula: C₁₀H₈
MW: 128.17 g/mol
Boiling Pt: 218 °C (1013 hPa)
Melting Pt: 79…82 °C
Density: 1.145 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00001742
CAS Number: 91-20-3
EINECS: 202-049-5
UN: 1334
ADR: 4.1,III
Merck Index: 12,06457
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c87c606cb828e0f39bab8b9d50c2c26501e1637d&vid=9e4bc21353014391fc8486208c8757e6375e2ef5&prid=web&clid=SAPDAM