2-Isopropylnaphthalene 96%
Supplier: Thermo Fisher Scientific
A15153.14 A15153.06
A15153.14EA 462 EUR
A15153.14 A15153.06
2-Isopropylnaphthalene 96%
2-Isopropylnaphthalene
Purity: 96% CAS No.: 2027-17-0 Molecular Formula: C13H14 Formula Weight: 170.26 Flash Point: 122°(252°F) Refractive Index: 1.5870 Melting Point: 14° Boiling Point: 268° MDL No.: MFCD00014322
Formula: C₁₃H₁₄
Boiling Pt: 268 °C
Melting Pt: 14 °C
Density: 0.975
Flash Pt: 122 °C (252 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014322
CAS Number: 2027-17-0
EINECS: 217-976-0
UN: 3077
ADR: 9,III